N-(2-Hydroxyethyl)iminodiacetic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-((2-Hydroxyethyl)azanediyl)diacetic acid
A228742-100G
A228742-25G
77667-900EA
125
USD
77667-900
77667-902
N-(2-Hydroxyethyl)iminodiacetic acid ≥98%
N-(2-Hydroxyethyl)iminodiacetic acid
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₅ MW: 177.155 g/mol |
MDL Number:
MFCD00004293 CAS Number: 93-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...