DL-Threonine methyl ester hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-Thr-OMe·HCl, Methyl threoninate hydrochloride
A228650-25G
A228650-5G
A228650-100G
77612-218EA
59.74
USD
77612-218
77612-220
77683-666
DL-Threonine methyl ester hydrochloride ≥98%
DL-Threonine methyl ester hydrochloride
Precaución: For research use only.
Formula:
C₅H₁₁NO₃·ClH MW: 169.607 g/mol |
MDL Number:
MFCD00070400 CAS Number: 62076-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...