1-Boc-5-cyanoindole-2-boronic acid ≥98%

Supplier: Ambeed
Warning

A228205-1G A228205-5G
77872-161EA 43 USD
77872-161 77872-162
1-Boc-5-cyanoindole-2-boronic acid ≥98%
1-Boc-5-cyanoindole-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BN₂O₄
MDL Number: MFCD04114578
CAS Number: 475102-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR