2-(4-Methylpiperazino)pyrimidine-5-boronic acid ≥98%

Supplier: Ambeed
Warning

A227570-100MG A227570-1G A227570-250MG
77885-568EA 46 USD
77885-568 77885-569 77885-570
2-(4-Methylpiperazino)pyrimidine-5-boronic acid ≥98%

Precaución: for Research Use Only

Formula: C₉H₁₅BN₄O₂
MDL Number: MFCD07375170
CAS Number: 1256355-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR