Cytidine, N-benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-2'-deoxy-5-methyl-, 3'-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite] ≥95%

Supplier: Ambeed
Warning

Synonyms: DMT-5Me-dC(Bz)-CE Phosphoramidite

A227478-1G A227478-250MG A227478-25G A227478-5G
77748-379EA 67 USD
77748-379 77748-380 77748-381 77748-382
Cytidine, N-benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-2'-deoxy-5-methyl-, 3'-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite] ≥95%
Cytidine, N-benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-2'-deoxy-5-methyl-, 3'-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Precaución: for Research Use Only

Formula: C₄₇H₅₄N₅O₈P
MW: 847.93 g/mol
MDL Number: MFCD00148939
CAS Number: 105931-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR