1-Benzyl-D-aspartate ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Amino-4-(benzyloxy)-4-oxobutanoic acid, H-D-Asp-OBzl, D-Aspartic Acid 1-Benzyl Ester

A226959-5G A226959-1G
77685-621EA 22 USD
77685-621 77902-062
1-Benzyl-D-aspartate ≥98%
1-Benzyl-D-aspartate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD00037289
CAS Number: 79337-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR