(S)-2-Amino-4-methyl-4-(3-(4-((S)-1-((S)-1-methylpyrrolidin-2-yl)ethoxy)pyrimidin-2-yl)-1,2,4-oxadiazol-5-yl)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile ≥99%

Supplier: Ambeed
Warning

Synonyms: BI-2865

A2261917-1MG A2261917-5MG
77626-156EA 210.12 USD
77626-156 77626-158
(S)-2-Amino-4-methyl-4-(3-(4-((S)-1-((S)-1-methylpyrrolidin-2-yl)ethoxy)pyrimidin-2-yl)-1,2,4-oxadiazol-5-yl)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile ≥99%
(S)-2-Amino-4-methyl-4-(3-(4-((S)-1-((S)-1-methylpyrrolidin-2-yl)ethoxy)pyrimidin-2-yl)-1,2,4-oxadiazol-5-yl)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile

Precaución: for Research Use Only

Formula: C₂₃H₂₇N₇O₂S
MW: 465.57 g/mol
CAS Number: 2937327-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR