2-Fluoro-5-(trifluoromethyl)pyridin-3-ol ≥97%

Supplier: Ambeed
Danger

A226027-5G A226027-1G
77615-884EA 1220.55 USD
77615-884 77748-372
2-Fluoro-5-(trifluoromethyl)pyridin-3-ol ≥97%
2-Fluoro-5-(trifluoromethyl)pyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₃F₄NO
MW: 181.0877 g/mol
MDL Number: MFCD16611121
CAS Number: 1227579-54-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR