(S)-2-(4-(2-(2-Methylazetidin-1-yl)-6-(trifluoromethyl)pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(piperazin-1-yl)ethan-1-one 98% 99.9+% ee

Supplier: Ambeed
Warning

A2259954-10MG A2259954-1MG A2259954-5MG
77598-336EA 512.5 USD
77598-336 77598-338 77598-340
(S)-2-(4-(2-(2-Methylazetidin-1-yl)-6-(trifluoromethyl)pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(piperazin-1-yl)ethan-1-one 98% 99.9+% ee
(S)-2-(4-(2-(2-Methylazetidin-1-yl)-6-(trifluoromethyl)pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(piperazin-1-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₈H₂₂F₃N₇O
CAS Number: 2568608-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR