2-Bromo-6-(1H-imidazol-1-yl)pyridine 95+%

Supplier: Ambeed
Warning

A225930-100MG A225930-1G A225930-250MG
77890-334EA 9 USD
77890-334 77890-335 77890-336
2-Bromo-6-(1H-imidazol-1-yl)pyridine 95+%
2-Bromo-6-(1H-imidazol-1-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₆BrN₃
MW: 224.06 g/mol
MDL Number: MFCD08668593
CAS Number: 463336-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR