3-(Iodomethyl)oxetane ≥98%

Supplier: Ambeed
Warning

A225770-100MG A225770-1G A225770-5G
77890-331EA 67 USD
77890-331 77890-332 77890-333
3-(Iodomethyl)oxetane ≥98%
3-(Iodomethyl)oxetane

Precaución: for Research Use Only

Formula: C₄H₇IO
MDL Number: MFCD18909284
CAS Number: 1003013-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR