LX-4211 ≥98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(methylthio)tetrahydro-2H-pyran-3,4,5-triol, Sotagliflozin

A225622-100MG A225622-10MG A225622-250MG A225622-5G A225622-1G A225622-50MG A225622-10G
77612-192EA 12.36 USD
77612-192 77612-194 77612-196 77612-198 77685-615 77685-616 77892-360
LX-4211 ≥98%
LX-4211

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClO₅S
MW: 424.94 g/mol
MDL Number: MFCD22493506
CAS Number: 1018899-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR