5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-1-(piperidin-4-yl)-1H-benzo[d]imidazol-2(3H)-one, 5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone

A225109-5G A225109-100G A225109-25G
77626-144EA 62.83 USD
77626-144 77877-708 77877-709
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥95%
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClN₃O
MDL Number: MFCD02093793
CAS Number: 53786-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR