N-(6-((1H-Pyrazol-1-yl)methyl)-4-methoxybenzo[d]isoxazol-3-yl)-2-methoxybenzenesulfonamide ≥98%

Supplier: Ambeed
Warning

A2250228-1MG A2250228-10MG A2250228-5MG
77598-334EA 76.88 USD
77598-334 77626-140 77626-142
N-(6-((1H-Pyrazol-1-yl)methyl)-4-methoxybenzo[d]isoxazol-3-yl)-2-methoxybenzenesulfonamide ≥98%
N-(6-((1H-Pyrazol-1-yl)methyl)-4-methoxybenzo[d]isoxazol-3-yl)-2-methoxybenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₄O₅S
MW: 414.44 g/mol
CAS Number: 2569008-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR