Phenyl methanesulfonate ≥97%

Supplier: Ambeed
Warning

Synonyms: Methanesulphonic acid Phenyl Ester

A224820-1G A224820-5G
77600-806EA 6.15 USD
77600-806 77667-872
Phenyl methanesulfonate ≥97%
Phenyl methanesulfonate

Precaución: for Research Use Only

Formula: C₇H₈O₃S
MW: 172.20 g/mol
MDL Number: MFCD00095143
CAS Number: 16156-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR