3,4-Dihydro-2H-chromen-3-ylamine hydrochloride ≥95%

Supplier: Ambeed
Warning

A224436-1G A224436-100MG A224436-250MG
77685-608EA 227 USD
77685-608 77901-467 77901-230
3,4-Dihydro-2H-chromen-3-ylamine hydrochloride ≥95%
3,4-Dihydro-2H-chromen-3-ylamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁NO·ClH
MDL Number: MFCD01736331
CAS Number: 18518-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR