5-Nitro-2-(trifluoromethoxy)aniline ≥98%

Supplier: Ambeed
Warning

A224174-1G A224174-10G
77671-980EA 35 USD
77671-980 77881-569
5-Nitro-2-(trifluoromethoxy)aniline ≥98%
5-Nitro-2-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O₃
MW: 222.12 g/mol
MDL Number: MFCD06248592
CAS Number: 158579-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR