5,5'-sulfonylbis(1,3-dibromo-2-(2,3-dibromopropoxy)benzene) 95+%

Supplier: Ambeed
Warning

A224074-100MG A224074-1G
77667-858EA 54 USD
77667-858 77667-860
5,5'-sulfonylbis(1,3-dibromo-2-(2,3-dibromopropoxy)benzene) 95+%
5,5'-sulfonylbis(1,3-dibromo-2-(2,3-dibromopropoxy)benzene)

Precaución: for Research Use Only

Formula: C₁₈H₁₄Br₈O₄S
MW: 965.59 g/mol
MDL Number: MFCD00329083
CAS Number: 42757-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR