Methyl cholate ≥95%

Supplier: Ambeed
Warning

Synonyms: 3α,7α,12α-Trihydroxycholan-24-oic acid methyl ester, Cholic acid methyl ester, 3α,7α,12α-Trihydroxy-5β-cholanic acid methyl ester, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid methyl ester, 3α,7α,12α-Trihydroxycholanic acid methyl ester, (R)-Methyl 4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoate

A223721-1G A223721-25G A223721-100MG A223721-5G
77626-130EA 17.51 USD
77626-130 77626-132 77879-359 77879-360
Methyl cholate ≥95%
Methyl cholate

Precaución: for Research Use Only

Formula: C₂₅H₄₂O₅
MW: 422.61
MDL Number: MFCD00064934
CAS Number: 1448-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR