4-Propylbenzene-1,3-diol ≥97%

Supplier: Ambeed
Warning

A223573-100MG A223573-250MG
77626-126EA 10.3 USD
77626-126 77626-128
4-Propylbenzene-1,3-diol ≥97%
4-Propylbenzene-1,3-diol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00020120
CAS Number: 18979-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR