(S)-2-((2-Amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl 2-(((benzyloxy)carbonyl)amino)-3-methylbutanoate ≥95%

Supplier: Ambeed
Warning

A223276-250MG A223276-1G A223276-25G A223276-5G
77612-170EA 9.27 USD
77612-170 77685-602 77685-603 77685-604
(S)-2-((2-Amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl 2-(((benzyloxy)carbonyl)amino)-3-methylbutanoate ≥95%
(S)-2-((2-Amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl 2-(((benzyloxy)carbonyl)amino)-3-methylbutanoate

Precaución: for Research Use Only

Formula: C₂₁H₂₆N₆O₆
MDL Number: MFCD08273968
CAS Number: 124832-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR