1-Bromo-2-fluoro-4,5-dimethylbenzene ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A223166-1G A223166-250MG
77683-641EA 53 USD
77683-641 77892-354
1-Bromo-2-fluoro-4,5-dimethylbenzene ≥95%, Ambeed.Inc
1-Bromo-2-fluoro-4,5-dimethylbenzene

Precaución: For research use only.

Formula: C₈H₈BrF
MDL Number: MFCD08695022
CAS Number: 5100-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR