Trisodium citrate ≥98%

Supplier: Ambeed
Warning

Synonyms: Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate

A223160-100G A223160-25G A223160-500G
77598-328EA 7.18 USD
77598-328 77598-330 77598-332
Trisodium citrate ≥98%
Trisodium citrate

Precaución: for Research Use Only

Formula: C₆H₅Na₃O₇
MW: 258.07 g/mol
MDL Number: MFCD00012462
CAS Number: 68-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR