Chlorotrispyrrolidinophosphonium hexafluorophosphate ≥96%

Supplier: Ambeed
Danger

Synonyms: Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, Chlorotripyrrolidinophosphonium hexafluorophosphate

A222809-5G A222809-25G
77593-446EA 17.43 USD
77593-446 77881-564
Chlorotrispyrrolidinophosphonium hexafluorophosphate ≥96%
Chloro-tris-pyrrolidinophosphonium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₂H₂₄ClF₆N₃P₂
MDL Number: MFCD00210035
CAS Number: 133894-48-1
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR