5-Cyano-5-nitrophenylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Nitro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

A222417-1G A222417-250MG
77881-559EA 240 USD
77881-559 77881-560
5-Cyano-5-nitrophenylboronic acid pinacol ester ≥98%
5-Cyano-5-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BN₂O₄
MDL Number: MFCD12546471
CAS Number: 1218791-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR