Polyethylene glycol mono-4-nonylphenyl ether ≥98% (by NMR)

Supplier: Ambeed
Warning

Synonyms: 4-Nonylphenol polyethoxylate

A222389-500G A222389-1000G A222389-100G
77600-780EA 75.85 USD
77600-780 77667-850 77667-852
Polyethylene glycol mono-4-nonylphenyl ether ≥98% (by NMR)
Polyethylene glycol mono-4-nonylphenyl ether

Precaución: for Research Use Only

MDL Number: MFCD00148006
CAS Number: 26027-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR