3-Aminocyclopentanol ≥98%

Supplier: Ambeed
Danger

A222268-100MG A222268-250MG
77671-970EA 66 USD
77671-970 77671-972
3-Aminocyclopentanol ≥98%
3-Aminocyclopentanol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD14705160
CAS Number: 13725-38-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR