2-(1H-Pyrazol-1-yl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 95+%

Supplier: Ambeed
Warning

A221408-1G A221408-250MG A221408-5G
77885-547EA 219 USD
77885-547 77885-548 77885-549
2-(1H-Pyrazol-1-yl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 95+%
2-(1H-Pyrazol-1-yl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₈BN₃O₂
MDL Number: MFCD09607672
CAS Number: 1186115-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR