6-Nitro-2,3,5-trifluoroaniline ≥95%

Supplier: Ambeed
Danger

A221362-100MG A221362-1G A221362-250MG
77881-552EA 59 USD
77881-552 77881-553 77881-554
6-Nitro-2,3,5-trifluoroaniline ≥95%
6-Nitro-2,3,5-trifluoroaniline

Precaución: for Research Use Only

Formula: C₆H₃F₃N₂O₂
MDL Number: MFCD00971838
CAS Number: 5415-62-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR