6-Iodo-o-toluic acid ≥97%

Supplier: Ambeed
Warning

A221120-1G A221120-100MG A221120-10G A221120-250MG
77671-962EA 52 USD
77671-962 77881-546 77881-547 77881-548
6-Iodo-o-toluic acid ≥97%
6-Iodo-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MDL Number: MFCD01310823
CAS Number: 54811-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR