Piperacillin ≥98%

Supplier: Ambeed
Danger

Synonyms: (2S,5R,6R)-6-((R)-2-(4-Ethyl-2,3-Dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

A220109-25G A220109-5G A220109-100G A220109-1G A220109-250MG
77612-142EA 63.86 USD
77612-142 77612-144 77612-136 77612-138 77612-140
Piperacillin ≥98%
Piperacillin

Precaución: for Research Use Only

Formula: C₂₃H₂₇N₅O₇S
MW: 517.557 g/mol
MDL Number: MFCD00865043
CAS Number: 61477-96-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR