Dichloromethyloctylsilane ≥97%

Supplier: Ambeed
Danger

A219567-25G A219567-5G
77626-110EA 26.78 USD
77626-110 77879-343
Dichloromethyloctylsilane ≥97%
Dichloromethyloctylsilane

Precaución: for Research Use Only

Formula: C₉H₂₀Cl₂Si
MDL Number: MFCD00042606
CAS Number: 14799-93-0
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR