4-Allylveratrole ≥95%

Supplier: Ambeed
Warning

Synonyms: 4-Allylveratrole, Eugenol methyll ether, 4-Prop-2-enylveratrole, 4-Allyl-1,2-Dimethoxybenzene, 4-Allyl-1,2-dimethoxybenzene, Methyl eugenol, Eugenol methyl ether, Methyleugenol

A218368-100G A218368-25G A218368-500G
77600-758EA 36.9 USD
77600-758 77600-760 77667-824
4-Allylveratrole ≥95%
4-Allylveratrole

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00008652
CAS Number: 93-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR