6-Bromo-8-fluoroisoquinoline ≥98%

Supplier: Ambeed
Warning

A218125-1G A218125-100MG A218125-250MG
77685-590EA 133 USD
77685-590 77892-345 77892-346
6-Bromo-8-fluoroisoquinoline ≥98%
6-Bromo-8-fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MDL Number: MFCD28125257
CAS Number: 1258833-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR