(3R,4R,5S)-4-Acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-enecarboxylic acid hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: Oseltamivir acid hydrochloride

A217687-1MG A217687-5MG
77615-824EA 16.48 USD
77615-824 77746-301
(3R,4R,5S)-4-Acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-enecarboxylic acid hydrochloride ≥97%
(3R,4R,5S)-4-Acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-enecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₅ClN₂O₄
MW: 320.81 g/mol
CAS Number: 1415963-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR