7-Nitrobenzo[d]thiazol-2-amine ≥95%

Supplier: Ambeed
Warning

A217645-1G A217645-100MG A217645-250MG
77671-944EA 125 USD
77671-944 77883-543 77883-544
7-Nitrobenzo[d]thiazol-2-amine ≥95%
7-Nitrobenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂S
MDL Number: MFCD01237562
CAS Number: 89793-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR