9H-Carbazole-3-carbonitrile ≥95%

Supplier: Ambeed
Warning

A217298-1G A217298-250MG A217298-5G
77593-414EA 36.9 USD
77593-414 77615-820 77685-586
9H-Carbazole-3-carbonitrile ≥95%
9H-Carbazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₃H₈N₂
MDL Number: MFCD00454925
CAS Number: 57102-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR