2-Amino-5-bromo-4-hydroxy-6-phenylpyrimidine ≥98%

Supplier: Ambeed
Warning

A216989-5G A216989-1G
77626-094EA 516 USD
77626-094 77879-330
2-Amino-5-bromo-4-hydroxy-6-phenylpyrimidine ≥98%
2-Amino-5-bromo-4-hydroxy-6-phenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₈BrN₃O
MDL Number: MFCD00813905
CAS Number: 56741-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR