Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate ≥98%

Supplier: Ambeed
Warning

Synonyms: Potassium (2-(((benzyloxy)carbonyl)amino)ethyl)trifluoroborate, Potassium (2-{[(benzyloxy)carbonyl]amino}ethyl)(trifluoro)borate

A216753-1G A216753-250MG
77872-104EA 47 USD
77872-104 77872-105
Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate ≥98%
Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₂BF₃NO₂·K
MW: 285.112 g/mol
MDL Number: MFCD10698612
CAS Number: 926280-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR