(S)-Piperazin-2-ylmethanol dihydrochloride ≥96%

Supplier: Ambeed

A216154-100MG A216154-1G A216154-250MG
77881-531EA 13 USD
77881-531 77881-532 77881-533
(S)-Piperazin-2-ylmethanol dihydrochloride ≥96%
(S)-Piperazin-2-ylmethanol dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₂O
MW: 189.08 g/mol
MDL Number: MFCD09954307
CAS Number: 149629-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR