3-Bromo-2-vinylpyridine ≥95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with TBC

A216097-1G A216097-250MG
77612-120EA 568.56 USD
77612-120 77885-534
3-Bromo-2-vinylpyridine ≥95% stabilized
3-Bromo-2-vinylpyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN
MDL Number: MFCD23135675
CAS Number: 799246-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR