2-Chloro-3-methoxy-6-(trifluoromethyl)pyridine ≥97%

Supplier: Ambeed
Warning

A215312-100MG A215312-250MG
77872-100EA 13 USD
77872-100 77872-101
2-Chloro-3-methoxy-6-(trifluoromethyl)pyridine ≥97%
2-Chloro-3-methoxy-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MDL Number: MFCD15478057
CAS Number: 1214348-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR