tert-Butyl 3-(dimethylamino)propanoate ≥95%

Supplier: Ambeed
Warning

A214509-1G A214509-250MG
77598-324EA 189.63 USD
77598-324 77748-336
tert-Butyl 3-(dimethylamino)propanoate ≥95%
tert-Butyl 3-(dimethylamino)propanoate

Precaución: for Research Use Only

Formula: C₉H₁₉NO₂
MW: 173.2527 g/mol
MDL Number: MFCD31699124
CAS Number: 88722-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR