2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-8-(tert-butoxycarbonyl)-2,8-diazaspiro[4.5]decane-3-carboxylic acid

A213727-100MG A213727-1G A213727-250MG
77872-088EA 49 USD
77872-088 77872-089 77872-090
2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester ≥95%
2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester

Precaución: for Research Use Only

Formula: C₂₉H₃₄N₂O₆
MW: 506.59 g/mol
MDL Number: MFCD07779072
CAS Number: 849928-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR