4-(3-Oxobutanoyl)benzonitrile ≥95%

Supplier: Ambeed
Warning

A213617-100MG A213617-250MG
77685-574EA 201 USD
77685-574 77685-575
4-(3-Oxobutanoyl)benzonitrile ≥95%
4-(3-Oxobutanoyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MDL Number: MFCD00477607
CAS Number: 62585-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR