(4-Ethoxy-2-formylphenyl)boronic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 4-Ethoxy-2-formylphenylboronic acid

A213606-1G A213606-250MG A213606-5G
77667-792EA 22 USD
77667-792 77872-084 77872-085
(4-Ethoxy-2-formylphenyl)boronic acid 95+%
(4-Ethoxy-2-formylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MDL Number: MFCD08458481
CAS Number: 1106867-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR