Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate ≥95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,4,4,4',4',4'-hexafluoro-DL-valinate, 4,4,4,4',4',4'-Hexafluorovaline, ethyl ester

A213594-250MG A213594-100MG A213594-1G
77612-102EA 41.2 USD
77612-102 77685-572 77685-573
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate ≥95%
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate

Precaución: for Research Use Only

Formula: C₇H₉F₆NO₂
MDL Number: MFCD00040845
CAS Number: 78185-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR