tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate ≥95%

Supplier: Ambeed
Danger

A213513-250MG A213513-25G
77890-269EA 7 USD
77890-269 77890-270
tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate ≥95%
tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD01317793
CAS Number: 177911-87-4
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR