2,6-Difluorophenylhydrazine hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Difluoro-2-hydrazinobenzene hydrochloride
A213336-1G
A213336-5G
77748-329EA
10
USD
77748-329
77748-330
2,6-Difluorophenylhydrazine hydrochloride ≥98%
2,6-Difluorophenylhydrazine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₆F₂N₂·ClH MW: 180.583 g/mol |
MDL Number:
MFCD03094169 CAS Number: 502496-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...