2-Bromo-4-fluoro-5-methylpyridine ≥95%

Supplier: Ambeed
Warning

A212843-100MG A212843-1G
77883-515EA 55 USD
77883-515 77883-516
2-Bromo-4-fluoro-5-methylpyridine ≥95%
2-Bromo-4-fluoro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD16609962
CAS Number: 1211537-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR